Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Bcl11b
Entrez Gene ID
100768427
Gene Name
B-cell CLL/lymphoma 11B (zinc finger protein)
Gene Aliases
I79_010590
Chromosome Location
Chr.Un|NW_003613993: 221996 - 294841 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003503713.2 XP_003503761.1 3 - 4 698 77

View Full Product Details