Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gng10
Entrez Gene ID
100755995
Gene Name
guanine nucleotide binding protein (G protein), gamma 10
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614906: 56795 - 59890 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003510573.2 XP_003510621.2 1 - 2 260 70

View Full Product Details