Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rtp4
Entrez Gene ID
100755040
Gene Name
receptor (chemosensory) transporter protein 4
Gene Aliases
I79_000870
Chromosome Location
Chr.Un|NW_003613596: 4712682 - 4717238 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007642600.1 XP_007640790.1 1 - 2 213 86

View Full Product Details