Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fdxacb1
Entrez Gene ID
100771758
Gene Name
ferredoxin-fold anticodon binding domain containing 1
Gene Aliases
I79_004382
Chromosome Location
Chr.Un|NW_003613694: 1110722 - 1117858 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003498525.2 XP_003498573.2 4 - 5 2345 59

View Full Product Details