Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ntsr2
Entrez Gene ID
100765229
Gene Name
neurotensin receptor 2
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613839: 758046 - 765327 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003501236.1 XP_003501284.1 3 - 4 984 84

View Full Product Details