Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tspo2
Entrez Gene ID
100756965
Gene Name
translocator protein 2
Gene Aliases
I79_020368
Chromosome Location
Chr.Un|NW_003615227: 194184 - 195493 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003511762.1 XP_003511810.1 1 - 2 175 63

View Full Product Details