Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dnajb4
Entrez Gene ID
100757372
Gene Name
DnaJ (Hsp40) homolog, subfamily B, member 4
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613622: 1873154 - 1883616 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003496433.2 XP_003496481.1 1 - 2 694 106
XM_007650117.1 XP_007648307.1 2 - 3 1059 106
GenBank mRNA JI874889.1 - 1 - 2 245 106
JP058887.1 - 1 - 2 245 106

View Full Product Details