Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Erp27
Entrez Gene ID
100754205
Gene Name
endoplasmic reticulum protein 27
Gene Aliases
I79_005249
Chromosome Location
Chr.Un|NW_003613723: 2257951 - 2275976 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003499208.2 XP_003499256.1 5 - 6 574 110

View Full Product Details