Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Igfbp1
Entrez Gene ID
100763682
Gene Name
insulin-like growth factor binding protein 1
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614057: 396920 - 400841 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003504385.1 XP_003504433.1 3 - 4 689 78

View Full Product Details