Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Bmp15
Entrez Gene ID
100756159
Gene Name
bone morphogenetic protein 15
Gene Aliases
I79_013065
Chromosome Location
Chr.Un|NW_003614190: 983542 - 988791 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003505803.2 XP_003505851.2 1 - 2 328 103

View Full Product Details