Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Kcnb2
Entrez Gene ID
100764283
Gene Name
potassium voltage-gated channel, Shab-related subfamily, member 2
Gene Aliases
I79_015548
Chromosome Location
Chr.Un|NW_003614432: 189818 - 372578 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003507806.2 XP_003507854.1 1 - 2 251 80

View Full Product Details