Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Otp
Entrez Gene ID
100760966
Gene Name
orthopedia homeobox
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613700: 1308349 - 1317999 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007632179.1 XP_007630369.1 3 - 4 491 65

View Full Product Details