Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slfn5
Entrez Gene ID
100750338
Gene Name
schlafen family member 5
Gene Aliases
I79_001089
Chromosome Location
Chr.Un|NW_003613601: 4595510 - 4603667 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003495826.1 XP_003495874.1 1 - 2 1000 78

View Full Product Details