Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ston1
Entrez Gene ID
100769184
Gene Name
stonin 1
Gene Aliases
Stonin-1
Chromosome Location
Chr.Un|NW_003613744: 970066 - 1008793 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003499539.2 XP_003499587.1 2 - 3 2059 104
XM_007640305.1 XP_007638495.1 3 - 4 2218 104

View Full Product Details