Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cdrt1
Entrez Gene ID
100761995
Gene Name
CMT1A duplicated region transcript 1
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613619: 2476989 - 2506759 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003496368.1 XP_003496416.1 5 - 6 1150 72

View Full Product Details