Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Spert
Entrez Gene ID
100752719
Gene Name
spermatid associated
Gene Aliases
I79_006734
Chromosome Location
Chr.Un|NW_003613795: 1641912 - 1651371 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003500472.1 XP_003500520.1 2 - 3 160 99

View Full Product Details