Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ccl19
Entrez Gene ID
100751942
Gene Name
chemokine (C-C motif) ligand 19
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614108: 1004619 - 1006118 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003504995.1 XP_003505043.1 2 - 3 193 78

View Full Product Details