Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Galp
Entrez Gene ID
100764710
Gene Name
galanin-like peptide
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003615631: 32081 - 55358 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007653274.1 XP_007651464.1 3 - 4 295 69

View Full Product Details