Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Areg
Entrez Gene ID
100772017
Gene Name
amphiregulin
Gene Aliases
I79_021127
Chromosome Location
Chr.Un|NW_003615432: 123881 - 164717 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007652867.1 XP_007651057.1 8 - 9 1506 94

View Full Product Details