Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mafk
Entrez Gene ID
100763840
Gene Name
v-maf avian musculoaponeurotic fibrosarcoma oncogene homolog K
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613618: 3972472 - 3984888 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003496301.2 XP_003496349.1 2 - 3 117 87

View Full Product Details