Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Slc25a47
Entrez Gene ID
100771881
Gene Name
solute carrier family 25, member 47
Gene Aliases
I79_010605
Chromosome Location
Chr.Un|NW_003613993: 1108696 - 1116256 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007644286.1 XP_007642476.1 3 - 4 2039 58

View Full Product Details