Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Slc48a1
Entrez Gene ID
100767330
Gene Name
solute carrier family 48 (heme transporter), member 1
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003615717: 146776 - 149652 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007653431.1 XP_007651621.1 1 - 2 189 63

View Full Product Details