Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Spink8
Entrez Gene ID
100767379
Gene Name
serine peptidase inhibitor, Kazal type 8 (putative)
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614047: 347368 - 379013 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007644785.1 XP_007642975.1 4 - 5 416 104

View Full Product Details