Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Synpr
Entrez Gene ID
100752738
Gene Name
synaptoporin
Gene Aliases
I79_010540
Chromosome Location
Chr.Un|NW_003613987: 522074 - 862895 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007644238.1 XP_007642428.1 4 - 5 405 76

View Full Product Details