Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lrrn4
Entrez Gene ID
100772398
Gene Name
leucine rich repeat neuronal 4
Gene Aliases
I79_000447
Chromosome Location
Chr.Un|NW_003613587: 5543064 - 5552268 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003495258.2 XP_003495306.1 4 - 5 1021 91

View Full Product Details