Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Micalcl
Entrez Gene ID
100763317
Gene Name
MICAL C-terminal like
Gene Aliases
I79_014721
Chromosome Location
Chr.Un|NW_003614332: 723859 - 759948 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003507118.2 XP_003507166.1 6 - 7 1791 64

View Full Product Details