Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ly6d
Entrez Gene ID
100774259
Gene Name
lymphocyte antigen 6 complex, locus D
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003617406: 65010 - 66385 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003515141.2 XP_003515189.1 1 - 2 181 68

View Full Product Details