Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ttc9
Entrez Gene ID
100763223
Gene Name
tetratricopeptide repeat domain 9
Gene Aliases
I79_014267
Chromosome Location
Chr.Un|NW_003614290: 756629 - 794376 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007647326.1 XP_007645516.1 3 - 4 307 60

View Full Product Details