Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Atoh8
Entrez Gene ID
100751745
Gene Name
atonal homolog 8 (Drosophila)
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614172: 5976 - 10671 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007646134.1 XP_007644324.1 1 - 2 398 77

View Full Product Details