Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Plscr5
Entrez Gene ID
100762648
Gene Name
phospholipid scramblase family, member 5
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614375: 659324 - 674289 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003507385.1 XP_003507433.1 4 - 5 626 84

View Full Product Details