Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Nip7
Entrez Gene ID
100759635
Gene Name
NIP7, nucleolar pre-rRNA processing protein
Gene Aliases
I79_016682
Chromosome Location
Chr.Un|NW_003614572: 35638 - 37826 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003508682.2 XP_003508730.1 1 - 2 123 61
GenBank mRNA JI872140.1 - 1 - 2 118 61

View Full Product Details