Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tmed6
Entrez Gene ID
100760131
Gene Name
transmembrane emp24 protein transport domain containing 6
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614572: 39755 - 47707 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003508684.2 XP_003508732.1 1 - 2 890 86

View Full Product Details