Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Klhl28
Entrez Gene ID
100755905
Gene Name
kelch-like family member 28
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003615410: 143434 - 167453 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003512270.2 XP_003512318.1 3 - 4 1505 66
XM_007652785.1 XP_007650975.1 3 - 4 1420 66
GenBank mRNA JP056375.1 - 1 - 2 210 66

View Full Product Details