Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mfng
Entrez Gene ID
100762875
Gene Name
MFNG O-fucosylpeptide 3-beta-N-acetylglucosaminyltransferase
Gene Aliases
I79_024169
Chromosome Location
Chr.Un|NW_003617007: 61409 - 81529 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003514828.2 XP_003514876.1 7 - 8 900 73

View Full Product Details