Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Clrn2
Entrez Gene ID
100769210
Gene Name
clarin 2
Gene Aliases
Clarin-2
Chromosome Location
Chr.Un|NW_003614204: 739198 - 749461 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003505978.2 XP_003506026.1 2 - 3 436 81

View Full Product Details