Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cript
Entrez Gene ID
100764597
Gene Name
cysteine-rich PDZ-binding protein
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003615099: 190559 - 199402 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003511401.2 XP_003511449.1 3 - 4 201 116
GenBank mRNA JI889249.1 - 3 - 4 194 116
JP058223.1 - 3 - 4 194 116

View Full Product Details