Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Klk14
Entrez Gene ID
100764613
Gene Name
kallikrein-related peptidase 14
Gene Aliases
I79_023132
Chromosome Location
Chr.Un|NW_003616291: 117609 - 121074 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007654230.1 XP_007652420.1 1 - 2 41 103

View Full Product Details