Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Il36a
Entrez Gene ID
100757088
Gene Name
interleukin 36, alpha
Gene Aliases
I79_024304
Chromosome Location
Chr.Un|NW_003617125: 11406 - 13397 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007654963.1 XP_007653153.1 3 - 4 284 74

View Full Product Details