Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zic5
Entrez Gene ID
100767568
Gene Name
Zic family member 5
Gene Aliases
I79_009812
Chromosome Location
Chr.Un|NW_003613939: 941540 - 954165 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007643701.1 XP_007641891.1 3 - 4 490 66

View Full Product Details