Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ccdc51
Entrez Gene ID
100762437
Gene Name
coiled-coil domain containing 51
Gene Aliases
I79_011280
Chromosome Location
Chr.Un|NW_003614047: 269255 - 286904 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003504242.2 XP_003504290.2 2 - 3 376 74

View Full Product Details