Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tm4sf19
Entrez Gene ID
100761891
Gene Name
transmembrane 4 L six family member 19
Gene Aliases
I79_000636
Chromosome Location
Chr.Un|NW_003613591: 1947891 - 1950810 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003495429.2 XP_003495477.1 2 - 3 267 88

View Full Product Details