Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ube2n
Entrez Gene ID
100752830
Gene Name
ubiquitin-conjugating enzyme E2N
Gene Aliases
I79_013714
Chromosome Location
Chr.Un|NW_003614231: 556294 - 590300 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003506360.2 XP_003506408.1 3 - 4 457 130

View Full Product Details