Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pla2g4e
Entrez Gene ID
100770894
Gene Name
phospholipase A2, group IVE
Gene Aliases
I79_003808
Chromosome Location
Chr.Un|NW_003613677: 2132714 - 2213337 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007622134.1 XP_007620324.1 17 - 18 2032 73

View Full Product Details