Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Serpind1
Entrez Gene ID
100751445
Gene Name
serpin peptidase inhibitor, clade D (heparin cofactor), member 1
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613919: 1245961 - 1253123 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003502822.1 XP_003502870.1 3 - 4 1253 74

View Full Product Details