Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Efcab9
Entrez Gene ID
100772537
Gene Name
EF-hand calcium binding domain 9
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613815: 552654 - 559729 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003500828.1 XP_003500876.1 1 - 2 132 64

View Full Product Details