Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Chst3
Entrez Gene ID
100758880
Gene Name
carbohydrate (chondroitin 6) sulfotransferase 3
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614681: 320275 - 322710 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003509291.2 XP_003509339.1 1 - 2 181 90

View Full Product Details