Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pttg1ip
Entrez Gene ID
100756425
Gene Name
pituitary tumor-transforming 1 interacting protein
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613799: 1311909 - 1329562 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003500562.2 XP_003500610.1 3 - 4 346 90
GenBank mRNA JI894381.1 - 3 - 4 180 90
JP053506.1 - 3 - 4 180 90

View Full Product Details