Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Etaa1
Entrez Gene ID
100750682
Gene Name
Ewing tumor-associated antigen 1
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613893: 1404861 - 1419888 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007642998.1 XP_007641188.1 3 - 4 360 125

View Full Product Details