Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Qtrtd1
Entrez Gene ID
100774164
Gene Name
queuine tRNA-ribosyltransferase domain containing 1
Gene Aliases
I79_003547
Chromosome Location
Chr.Un|NW_003613667: 2110367 - 2135974 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003497882.2 XP_003497930.1 5 - 6 688 63
XM_007618907.1 XP_007617097.1 5 - 6 691 63
XM_007618916.1 XP_007617106.1 5 - 6 698 63
XM_007618931.1 XP_007617121.1 3 - 4 287 63

View Full Product Details