Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Qtrtd1
Entrez Gene ID
100774164
Gene Name
queuine tRNA-ribosyltransferase domain containing 1
Gene Aliases
I79_003547
Chromosome Location
Chr.Un|NW_003613667: 2110367 - 2135974 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003497882.2 XP_003497930.1 7 - 8 1101 55
XM_007618907.1 XP_007617097.1 7 - 8 1104 55
XM_007618916.1 XP_007617106.1 7 - 8 1111 55
XM_007618931.1 XP_007617121.1 5 - 6 700 55
GenBank mRNA JI895502.1 - 1 - 2 121 55

View Full Product Details